ENA390346766 Enamine

4-(aminomethyl)-2-bromo-6-methoxyphenol hydrochloride

  • MDL number MFCD23701789
  • Cas Number : 1432679-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123457
Availability details for
Quantity
  • Availability
  • Add to Cart