ENA390346771 Enamine

2-bromobicyclo[2.2.1]heptan-1-amine hydrochloride

  • MDL number MFCD23701795
  • Cas Number : 1432679-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124357
Availability details for
Quantity
  • Availability
  • Add to Cart