ENA390346774 Enamine

1-methyl-4,5-dihydro-1H-pyrazol-3-amine hydrochloride

  • MDL number MFCD23701797
  • Cas Number : 3956-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124589

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124589
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart