ENA390346776 Enamine

2-[(1H-1,2,3-triazol-1-yl)methyl]piperidine dihydrochloride

  • MDL number MFCD23701799
  • Cas Number : 1432679-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124672
Availability details for
Quantity
  • Availability
  • Add to Cart