ENA390346781 Enamine

1-(oxolan-3-yl)-1-phenylmethanamine hydrochloride

  • MDL number MFCD23701805
  • Cas Number : 1432678-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124821
Availability details for
Quantity
  • Availability
  • Add to Cart