ENA390346782 Enamine

3-[(oxolan-3-yl)methyl]piperidine hydrochloride

  • MDL number MFCD23701807
  • Cas Number : 1432679-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124863
Availability details for
Quantity
  • Availability
  • Add to Cart