ENA390346795 Enamine

1-(piperidine-1-carbonyl)-6-azaspiro[2.5]octane hydrochloride

  • MDL number MFCD23701822
  • Cas Number : 1432678-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125053
Availability details for
Quantity
  • Availability
  • Add to Cart