ENA390346801 Enamine

4-amino-6-methyl-1,2-dihydropyrimidin-2-one hydrochloride

  • MDL number MFCD23701828
  • Cas Number : 141625-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125112

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125112
Availability details for
Quantity
  • Availability
  • Add to Cart