ENA390346807 Enamine

4-amino-1-(2,6-difluorophenyl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD23701837
  • Cas Number : 1432680-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125174
Availability details for
Quantity
  • Availability
  • Add to Cart