ENA390346815 Enamine

3-{5H,6H,7H,8H-imidazo[1,2-a]pyrazin-7-yl}-2,2-dimethylpropanoic acid dihydrochloride

  • MDL number MFCD23701845
  • Cas Number : 1432678-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125287
Availability details for
Quantity
  • Availability
  • Add to Cart