ENA390346824 Enamine

6,7-dichloro-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD23701856
  • Cas Number : 1432680-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125415
Availability details for
Quantity
  • Availability
  • Add to Cart