ENA390346829 Enamine

diethyl(2-{[3-(trifluoromethyl)phenyl]amino}ethyl)amine dihydrochloride

  • MDL number MFCD23701863
  • Cas Number : 1432680-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125470

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125470
Availability details for
Quantity
  • Availability
  • Add to Cart