ENA390346830 Enamine

ethyl 2-(3-aminopropanamido)acetate hydrochloride

  • MDL number MFCD23701864
  • Cas Number : 92503-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125472
Availability details for
Quantity
  • Availability
  • Add to Cart