ENA390346832 Enamine

2-methyl-4-[4-(3-methylphenyl)piperazin-1-yl]butan-2-amine hydrochloride

  • MDL number MFCD23701866
  • Cas Number : 1432679-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125499
Availability details for
Quantity
  • Availability
  • Add to Cart