ENA390346847 Enamine

5-methyl-6-nitro-2,3-dihydro-1H-indole hydrochloride

  • MDL number MFCD23701881
  • Cas Number : 1432680-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125718

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125718
Availability details for
Quantity
  • Availability
  • Add to Cart