ENA390346848 Enamine

2-amino-1-methylcyclohexane-1-carboxylic acid hydrochloride

  • MDL number MFCD23701882
  • Cas Number : 1432680-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247...249
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125722

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125722
Availability details for
Quantity
  • Availability
  • Add to Cart