ENA390346853 Enamine

6-(3-ethylpiperidin-1-yl)-5-methylpyridin-3-amine hydrochloride

  • MDL number MFCD23701887
  • Cas Number : 1432678-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125772

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125772
Availability details for
Quantity
  • Availability
  • Add to Cart