ENA390346856 Enamine

3-{[4-(1,2,3,6-tetrahydropyridin-4-yl)phenoxy]methyl}benzonitrile hydrochloride

  • MDL number MFCD23701890
  • Cas Number : 1432680-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125786
Availability details for
Quantity
  • Availability
  • Add to Cart