ENA390346859 Enamine

1-[4-chloro-3-(trifluoromethyl)phenyl]-1-cyclopropylmethanamine hydrochloride

  • MDL number MFCD23701894
  • Cas Number : 1432679-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125827
Availability details for
Quantity
  • Availability
  • Add to Cart