ENA390346864 Enamine

5-(aminomethyl)-N,N-dimethyloxolane-2-carboxamide hydrochloride

  • MDL number MFCD23701901
  • Cas Number : 1432678-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125870
Availability details for
Quantity
  • Availability
  • Add to Cart