ENA390346868 Enamine

2-aminoethyl carbamate hydrochloride

  • MDL number MFCD23701905
  • Cas Number : 73711-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125908

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125908
Availability details for
Quantity
  • Availability
  • Add to Cart