ENA390346875 Enamine

2-bromo-5-methanesulfonyl-3-nitrothiophene

  • MDL number MFCD23701912
  • Cas Number : 2160-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125961

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125961
Availability details for
Quantity
  • Availability
  • Add to Cart