ENA390346880 Enamine

3-(2-chloroethyl)-8-oxa-3-azabicyclo[3.2.1]octane hydrochloride

  • MDL number MFCD23701918
  • Cas Number : 1432680-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126058
Availability details for
Quantity
  • Availability
  • Add to Cart