ENA390346881 Enamine

3-(5-bromopyridin-3-yl)propanoic acid hydrochloride

  • MDL number MFCD23701919
  • Cas Number : 1432681-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126060
Availability details for
Quantity
  • Availability
  • Add to Cart