ENA390346891 Enamine

2-{4-[2-(3,4-dimethylpyrrolidin-1-yl)ethoxy]phenyl}ethan-1-amine dihydrochloride

  • MDL number MFCD23701933
  • Cas Number : 1432681-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126179
Availability details for
Quantity
  • Availability
  • Add to Cart