ENA390346909 Enamine

tert-butyl 2-(3,4-dihydro-2H-pyran-5-yl)-2-oxoacetate

  • MDL number MFCD23701952
  • Cas Number : 1432680-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126363
Availability details for
Quantity
  • Availability
  • Add to Cart