ENA390346913 Enamine

4-(3-aminopropyl)-1$l^{6},4-thiomorpholine-1,1-dione hydrochloride

  • MDL number MFCD23701956
  • Cas Number : 1432677-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126408
Availability details for
Quantity
  • Availability
  • Add to Cart