ENA390346914 Enamine

3-(difluoromethyl)-4-methanesulfonylaniline hydrochloride

  • MDL number MFCD23701957
  • Cas Number : 1432678-69-7
  • Molecular Weight : 257.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(difluoromethyl)-4-(methylsulfonyl)aniline hydrochloride
Inchi Code 1S/C8H9F2NO2S.ClH/c1-14(12,13)7-3-2-5(11)4-6(7)8(9)10;/h2-4,8H,11H2,1H3;1H
Inchi Key PUHLFMAJQACXFX-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126409
Availability details for
Quantity
  • Availability
  • Add to Cart