ENA390346918 Enamine

2-amino-3-methoxypropanenitrile hydrochloride

  • MDL number MFCD23701963
  • Cas Number : 1432678-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126476

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126476
Availability details for
Quantity
  • Availability
  • Add to Cart