ENA390346922 Enamine

N-methyl-2-(propan-2-yl)-1,3-benzoxazol-5-amine dihydrochloride

  • MDL number MFCD23701967
  • Cas Number : 1432681-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126527

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126527
Availability details for
Quantity
  • Availability
  • Add to Cart