ENA390346924 Enamine

{1-azaspiro[3.5]nonan-7-yl}methanol

  • MDL number MFCD23701969
  • Cas Number : 1432680-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126537
Availability details for
Quantity
  • Availability
  • Add to Cart