ENA390346926 Enamine

[1-(4-hydrazinylbenzoyl)pyrrolidin-2-yl]methanol hydrochloride

  • MDL number MFCD23701972
  • Cas Number : 1432680-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126556
Availability details for
Quantity
  • Availability
  • Add to Cart