ENA390346928 Enamine

2-amino-4-methoxybutanamide hydrochloride

  • MDL number MFCD23701975
  • Cas Number : 1432677-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126565

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126565
Availability details for
Quantity
  • Availability
  • Add to Cart