ENA390346930 Enamine

2-(propan-2-yl)azetidine hydrochloride

  • MDL number MFCD23701977
  • Cas Number : 1432681-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126572
Availability details for
Quantity
  • Availability
  • Add to Cart