ENA390346931 Enamine

1,1-difluoro-4-phenylbutan-2-amine hydrochloride

  • MDL number MFCD23701978
  • Cas Number : 1432681-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126577
Availability details for
Quantity
  • Availability
  • Add to Cart