ENA390346932 Enamine

3-(pyrrolidin-1-yl)propanamide hydrochloride

  • MDL number MFCD23701979
  • Cas Number : 4996-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126578

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126578
Availability details for
Quantity
  • Availability
  • Add to Cart