ENA390346936 Enamine

4,5-dichloro-1,2,3,6-tetrahydropyridine

  • MDL number MFCD23701983
  • Cas Number : 1432680-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126628
Availability details for
Quantity
  • Availability
  • Add to Cart