ENA390346940 Enamine

6-(aminomethyl)-2-cyclopropylpyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD23701987
  • Cas Number : 1432678-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126662
Availability details for
Quantity
  • Availability
  • Add to Cart