ENA390346946 Enamine

6-(2,2-difluoroethoxy)pyridine-3-carboxylic acid

  • MDL number MFCD23701786
  • Cas Number : 1072855-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120587

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120587
Availability details for
Quantity
  • Availability
  • Add to Cart