ENA390346950 Enamine

1-(3,3-diethoxypropyl)pyrrolidine

  • MDL number MFCD00969490
  • Cas Number : 24299-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121069

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121069
Availability details for
Quantity
  • Availability
  • Add to Cart