ENA390346952 Enamine

5-phenylpyrrolidin-2-one

  • MDL number MFCD01687227
  • Cas Number : 22050-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-108
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121433
Availability details for
Quantity
  • Availability
  • Add to Cart