ENA390346958 Enamine

2-(cyclohexyloxy)pyridine-4-carboxylic acid

  • MDL number MFCD09258836
  • Cas Number : 1019353-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125221
Availability details for
Quantity
  • Availability
  • Add to Cart