ENA390346963 Enamine

2-[(4-methoxyphenyl)methoxy]ethan-1-amine

  • MDL number MFCD11147572
  • Cas Number : 40141-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125227
Availability details for
Quantity
  • Availability
  • Add to Cart