ENA390346964 Enamine

N-(prop-2-en-1-yl)methanesulfonamide

  • MDL number MFCD00465166
  • Cas Number : 59639-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125228
Availability details for
Quantity
  • Availability
  • Add to Cart