ENA390346976 Enamine

4-(acetyloxy)-3,3-dimethylbutanoic acid

  • MDL number MFCD20657194
  • Cas Number : 881019-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 75
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125242
Availability details for
Quantity
  • Availability
  • Add to Cart