ENA390346981 Enamine

1-[4-(chloromethyl)phenyl]-3-methyl-1H-pyrazole

  • MDL number MFCD16770030
  • Cas Number : 1248072-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125276
Availability details for
Quantity
  • Availability
  • Add to Cart