ENA390346982 Enamine

2-[4-(hydroxymethyl)phenyl]acetonitrile

  • MDL number MFCD12911541
  • Cas Number : 144825-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122773
Availability details for
Quantity
  • Availability
  • Add to Cart