ENA390346991 Enamine

2-(pyrrolidin-3-yloxy)-5-(trifluoromethyl)pyridine hydrochloride

  • MDL number MFCD13561268
  • Cas Number : 1220016-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125292
Availability details for
Quantity
  • Availability
  • Add to Cart