ENA390346994 Enamine

4H,5H,6H-cyclopenta[d][1,3]thiazole-2-carbaldehyde

  • MDL number MFCD10700212
  • Cas Number : 933728-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-125295
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart