ENA390346998 Enamine

4-ethyl-6-(methylsulfanyl)-1,3,5-triazin-2-amine

  • MDL number MFCD20706273
  • Cas Number : 23527-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125299

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125299
Availability details for
Quantity
  • Availability
  • Add to Cart